Learn More
MUCDHL, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MUCDHL.
Supplier: Abnova Corporation H00053841A01
Description
This gene is a novel mucin-like gene that is a member of the cadherin superfamily. While encoding nonpolymorphic tandem repeats rich in proline, serine and threonine similar to mucin proteins, the gene also contains sequence encoding calcium-binding motifs found in all cadherins. The role of the hybrid extracellular region and the specific function of this protein have not yet been determined. Alternative splicing has been identified, with observed variation resulting in the presence or absence of domains. In addition, splicing occurs in the 5' UTR but transcripts including these variations have not been described completely. [provided by RefSeq]
Sequence: AQYCSVNKDIFEVEENTNVTEPLVDIHVPEGQEVTLGALSTPFAFRIQGNQLFLNVTPDYEEKSLLEAQLLCQSGGTLVTQLRVFVSVLDVNDNAPEFPSpecifications
MUCDHL | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant MUCDHL. | |
MUPCDH | |
FLJ20219/MU-PCDH/MUCDHL | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_021924 | |
MUPCDH | |
MUCDHL (NP_068743, 27 a.a. ∽ 125 a.a) partial recombinant protein with GST tag. | |
RUO | |
53841 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only