Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MVP, Mouse, Clone: 1014, Abnova™

Catalog No. 89135510 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-510 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-510 Supplier Abnova Corporation Supplier No. MAB3767
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant MVP.

Specifications

Antigen MVP
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 1014
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant MVP.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from purified antibody
Host Species Mouse
Immunogen Recombinant protein corresponding to MVP.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific to a 104 KDa Major Vault Protein (MVP), also described as the lung resistance protein (LRP), and has shown to interact with the estrogen receptor. This antibody can be applied for the detection of MVP/LRP in a large number of eukaryotic cells including the MCF-7 and Hela tumor cell lines.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.