Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYC, Mouse, Clone: Myc.A7, Abnova™

Catalog No. 89136006 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-006 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-006 Supplier Abnova Corporation Supplier No. MAB5543
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of MYC.

Sequence: EQKLISEEDL

Specifications

Antigen MYC
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Myc.A7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of MYC.
Dilution Western Blot (1:1000) immunoprecipitation (1:200) Immunohistochemistry (1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.05% sodium azide)
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 410-419 of human MYC.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes overexpressed proteins containing MYC epitope tag fused to either amino- or carboxy-termini of targeted proteins in transfected mammalian cells.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.