Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYD88, Mouse, Clone: 2E9C2, Abnova™

Catalog No. 89111117 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-117 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-117 Supplier Abnova Corporation Supplier No. MAB2745
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant MYD88.

Specifications

Antigen MYD88
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2E9C2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant MYD88.
Dilution Western Blot (2-4 mg/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene MYD88
Gene Alias MYD88D
Gene Symbols MYD88
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 176-280 of human MYD88.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4615
Target Species Human, Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.