Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL2, Mouse, Clone: AT3B2, Abnova™

Catalog No. 89112336 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-336 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-336 Supplier Abnova Corporation Supplier No. MAB5641
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant MYL2.

Thus gene encodes the regulatory light chain associated with cardiac myosin beta (or slow) heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy. [provided by RefSeq]

Specifications

Antigen MYL2
Applications ELISA, Western Blot
Classification Monoclonal
Clone AT3B2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant MYL2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene MYL2
Gene Alias CMH10/DKFZp779C0562/MLC2
Gene Symbols MYL2
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human MYL2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4633
Target Species Human, Rat
Content And Storage Store at 4°C. For long term storage store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.