Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL3, Mouse, Clone: 3F8, Abnova™

Catalog No. 89108615 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-615 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-615 Supplier Abnova Corporation Supplier No. MAB10429
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant MYL3.

MYL3 encodes myosin light chain 3, an alkali light chain also referred to in the literature as both the ventricular isoform and the slow skeletal muscle isoform. Mutations in MYL3 have been identified as a cause of mid-left ventricular chamber type hypertrophic cardiomyopathy. [provided by RefSeq

Specifications

Antigen MYL3
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3F8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant MYL3.
Dilution Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation In ascitic (0.03% sodium azide)
Gene MYL3
Gene Alias CMH8/MLC1SB/MLC1V/VLC1
Gene Symbols MYL3
Host Species Mouse
Immunogen Recombinant protein corresponding to human MYL3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4634
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.