Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYLK2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MYLK2.
Supplier: Abnova Corporation H00085366A01
Description
This gene encodes a myosin light chain kinase, a calcium/calmodulin dependent enzyme, that is exclusively expressed in adult skeletal muscle. [provided by RefSeq
Sequence: KLLAKKPPSEASELTFEGVPMTHSPTDPRPAKAEEGKNILAESQKEVGEKTPGQAGQAKMQGDTSRGIEFQAVPSEKSEVGQALCLTAREEDCFQILDDCSpecifications
MYLK2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant MYLK2. | |
MYLK2 | |
KMLC/MLCK/MLCK2/skMLCK | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_033118 | |
MYLK2 | |
MYLK2 (NP_149109, 161 a.a. ∽ 260 a.a) partial recombinant protein with GST tag. | |
RUO | |
85366 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction