Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYO7A, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104970 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-970 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-970 Supplier Abnova Corporation Supplier No. H00004647A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant MYO7A.

This gene is a member of the myosin gene family. Myosins are mechanochemical proteins characterized by the presence of a motor domain, an actin-binding domain, a neck domain that interacts with other proteins, and a tail domain that serves as an anchor. This gene encodes an unconventional myosin with a very short tail. Defects in this gene are associated with the mouse shaker-1 phenotype and the human Usher syndrome 1B which are characterized by deafness, reduced vestibular function, and (in human) retinal degeneration. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Sequence: KQTTEPNFPEILLIAINKYGVSLIDPKTKDILTTHPFTKISNWSSGNTYFHITIGNLVRGSKLLCETSLGYKMDDLLTSYISQMLTAMSKQRGSRS

Specifications

Antigen MYO7A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant MYO7A.
Formulation 50% glycerol
Gene MYO7A
Gene Accession No. NM_000260
Gene Alias DFNA11/DFNB2/MYOVIIA/MYU7A/NSRD2/USH1B
Gene Symbols MYO7A
Host Species Mouse
Immunogen MYO7A (NP_000251, 2118 a.a. ∽ 2213 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4647
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.