Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYO9A, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104971 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-971 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-971 Supplier Abnova Corporation Supplier No. H00004649A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant MYO9A.

Sequence: GKRNIHRKTGHDDTAPCAILKSMDSFSFLQHPVHQRSLEILQRCKEEKYSITRKNPRTPLSDLQGMNALNEKNQHDTFDIAWNGRTGIRQSRLSSGTSLLDKDGIFANST

Specifications

Antigen MYO9A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant MYO9A.
Formulation 50% glycerol
Gene MYO9A
Gene Accession No. NM_006901
Gene Alias FLJ11061/FLJ13244/MGC71859
Gene Symbols MYO9A
Host Species Mouse
Immunogen MYO9A (NP_008832, 719 a.a. ∽ 828 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4649
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.