Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYOD1, Mouse, Clone: MYD712, Abnova™

Catalog No. 89110282 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-282 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-282 Supplier Abnova Corporation Supplier No. MAB12139

Mouse monoclonal antibody raised against recombinant human MYOD1.

This gene encodes a nuclear protein that belongs to the basic helix-loop-helix family of transcription factors and the myogenic factors subfamily. It regulates muscle cell differentiation by inducing cell cycle arrest, a prerequisite for myogenic initiation. The protein is also involved in muscle regeneration. It activates its own transcription which may stabilize commitment to myogenesis. [provided by RefSeq

Specifications

Antigen MYOD1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone MYD712
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant human MYOD1.
Dilution Immunohistochemistry (0.5-1 ug/mL) Western blot (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene MYOD1
Gene Alias MYF3/MYOD/PUM/bHLHc1
Gene Symbols MYOD1
Host Species Mouse
Immunogen Recombinant protein corresponding to human MYOD1.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4654
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.