Learn More
MYOG, Mouse, Clone: SPM144, Abnova™
Mouse monoclonal antibody raised against MYOG.
Supplier: Abnova Corporation MAB12116
Description
Myogenin is a muscle-specific transcription factor that can induce myogenesis in a variety of cell types in tissue culture. It is a member of a large family of proteins related by sequence homology, the helix-loop-helix (HLH) proteins. It is essential for the development of functional skeletal muscle. [provided by RefSeq
Specifications
MYOG | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (0.5-1 ug/mL) Western blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
MYOG | |
MYOG | |
Recombinant protein corresponding to human MYOG. | |
100 μg | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
SPM144 | |
Mouse monoclonal antibody raised against MYOG. | |
In PBS (0.05% BSA, 0.05% sodium azide) | |
MYF4/MYOGENIN/bHLHc3 | |
Mouse | |
Protein A/G purification | |
RUO | |
4656 | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.