Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin, Mouse, Clone: MY-32, Abnova™

Catalog No. 89135940 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-940 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-940 Supplier Abnova Corporation Supplier No. MAB5343

Mouse monoclonal antibody raised against native Myosin.

Specifications

Antigen Myosin
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone MY-32
Conjugate Unconjugated
Formulation In PBS (0.09% sodium azide, 1% BSA)
Host Species Mouse
Immunogen Native purified Myosin from rabbit skeletal muscle.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes myosin, the main constituent of the thick filaments of muscle fibres. Myosin acts with actin in the contraction and relaxation of muscles. This antibody detects skeletal muscle myosin specifically the fast twitch (type II) isomyosin molecules of the heavy chain of human or animal myosin.
Target Species Bovine, Chicken, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.