Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin, Mouse, Clone: MY-38, Abnova™
Mouse monoclonal antibody raised against native myosin.
Supplier: Abnova Corporation MAB1522
Specifications
Myosin | |
Monoclonal | |
Unconjugated | |
Western Blot (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
Mouse | |
Affinity purification | |
RUO | |
Human, Rabbit, Rat | |
Lyophilized |
Immunohistochemistry (PFA fixed), Western Blot | |
MY-32 | |
Mouse monoclonal antibody raised against native myosin. | |
Lyophilized from 1.2% sodium acetate (2mg BSA, 0.01mg sodium azide) | |
Native purified rabbit myosin. | |
100 μg | |
Primary | |
Store at -20°C on dry atmosphere. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction