Learn More
MYPN, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MYPN.
Supplier: Abnova Corporation H00084665A01
Description
MYPN is a component of the sarcomere that tethers nebulin (MIM 161650) in skeletal muscle and nebulette (MIM 605491) in cardiac muscle to alpha-actinin (see ACTN2; MIM 102573) at the Z lines (Bang et al., 2001 [PubMed 11309420]).[supplied by OMIM
Sequence: PDLSAFLSQEELDESVNLARLAINYDPLEKADETQARKRLSPDQMKHSPNLSFEPNFCQDNPRSPTSSKESPQEAKRPQYCSETQSKKVFLNKAADFIEELSSLFKSHSSSpecifications
MYPN | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant MYPN. | |
MYPN | |
MYOP | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_032578 | |
MYPN | |
MYPN (NP_115967, 61 a.a. ∽ 170 a.a) partial recombinant protein with GST tag. | |
RUO | |
84665 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only