Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-N6-methyladenosine (m6A), Clone: 17-3-4-1,

Catalog No. MABE1006MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE1006MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1006MI Supplier MilliporeSigma™ Supplier No. MABE1006
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects N6-methyladenosine (m6A) clone: 17-3-4-1 in All samples, and it is validated for Dot Blot, Immunoprecipitation

N6-Methyladenosine (m6A ) is an abundant modification found in mRNA, tRNA, snRNA, as well as long non-coding RNA, in all species. RNA adenosine methylation is catalyzed by a multicomponent complex composed of METTL3/MT-A70, METTL14, and WTAP in mammals. METTL3 & METTL14 are responsible for the methyltransferase activity of the complex, and WTAP mediates substrate recruitment. Fat mass and obesity-associated (FTO) and ALKBH5 are two identified demethylases that catalyze oxidative demthylation of m6A. Several functions of m6A modification have been proposed, including mRNA decay, splicing regulation, RNA nuclear export and maturation. (PMID 25171402). Anti-N6-methyladenosine (m6A), clone 17-3-4-1, is a mouse monoclonal that specifically detects RNA molecules with m6A modication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen N6-methyladenosine (m6A)
Applications Dot Blot, Immunoprecipitation
Classification Monoclonal
Clone 17-3-4-1
Formulation Purified mouse monoclonal IgG1λ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen Hapten N6-methyladenosine-5'-mono-phosphate conjugated to BSA of all N6-methyladenosine (m6A).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Target Species All species
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.