Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NACC1, Mouse, Clone: 6H2, Abnova™

Catalog No. 89109027 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-027 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-027 Supplier Abnova Corporation Supplier No. MAB10824
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant NACC1.

Members of the BTB/POZ family of transcriptional regulators, including BTBD14B, contain a conserved motif in the N-terminal region critical for protein-protein interactions and assembly of high molecular mass complexes (Korutla et al., 2002 [PubMed 11906783]).[supplied by OMIM

Specifications

Antigen NACC1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6H2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant NACC1.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene NACC1
Gene Alias BEND8/BTBD14B/FLJ37383/NAC-1/NAC1
Gene Symbols NACC1
Host Species Mouse
Immunogen Recombinant protein corresponding to human NACC1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112939
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.