Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAMPT, Mouse, Clone: 4D5, Abnova™

Catalog No. 89109390 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-390 Supplier Abnova Corporation Supplier No. MAB1122
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant NAMPT.

This gene encodes a protein that catalyzes the condensation of nicotinamide with 5-phosphoribosyl-1-pyrophosphate to yield nicotinamide mononucleotide, one step in the biosynthesis of nicotinamide adenine dinucleotide. The protein is an adipokine that is localized to the bloodstream and has various functions, including the promotion of vascular smooth muscle cell maturation and inhibition of neutrophil apoptosis. It also activates insulin receptor and has insulin-mimetic effects, lowering blood glucose and improving insulin sensitivity. The protein is highly expressed in visceral fat and serum levels of the protein correlate with obesity. This gene has a pseudogene on chromosome 10. [provided by RefSeq

Specifications

Antigen NAMPT
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant NAMPT.
Dilution Western Blot (1:1000-1:3000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene NAMPT
Gene Accession No. NM_005746
Gene Alias 1110035O14Rik/DKFZp666B131/MGC117256/PBEF/PBEF1/VF/VISFATIN
Gene Symbols NAMPT
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human NAMPT.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10135
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.