Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NANOG, Mouse, Clone: 5A10, Abnova™

Catalog No. 89110784 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-784 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-784 Supplier Abnova Corporation Supplier No. MAB2042
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant NANOG.

Specifications

Antigen NANOG
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5A10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant NANOG.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene NANOG
Gene Symbols NANOG
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-154 of human NANOG.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79923
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.