Learn More
NAT1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant NAT1.
Supplier: Abnova Corporation H00000009A01
Description
This gene is one of two arylamine N-acetyltransferase (NAT) genes in the human geneome, and is orthologous to the mouse and rat Nat2 genes. The enzyme encoded by this gene catalyzes the transfer of an acetyl group from acetyl-CoA to various arylamine and hydrazine substrates. This enzyme helps metabolize drugs and other xenobiotics, and functions in folate catabolism. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: DFESMNTYLQTSPSSVFTSKSFCSLQTPDGVHCLVGFTLTHRRFNYKDNTDLIEFKTLSEEEIEKVLKNIFNISLQRKLVPKHGDRFFTISpecifications
NAT1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant NAT1. | |
NAT1 | |
AAC1/NATI | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_000662 | |
NAT1 | |
NAT1 (NP_000653, 201 a.a. ∽ 290 a.a) partial recombinant protein with GST tag. | |
RUO | |
9 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only