Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Natural Killer Cell Activation Structures, Mouse, Clone: ANK61, Abnova™

Catalog No. 89135916 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-916 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-916 Supplier Abnova Corporation Supplier No. MAB5198
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Natural Killer Cell Activation Structures.

Specifications

Antigen Natural Killer Cell Activation Structures
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone ANK61
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Natural Killer Cell Activation Structures.
Dilution The optimal working dilution should be determined by the end user.
Formulation In culture supernatant (0.05% sodium azide)
Host Species Mouse
Immunogen Native purified from IL-2-activated cultured NK cells of Wag rats.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity ANK61 reactivity is rat strain-independent. The antigen recognized by ANK61 is highly expressed on freshly isolated and cultured rat NK cells. The antibodies also bind to rat alphabeta-TCR T cells and at a low level to rat B cells. Binding to other cell types is unknown. Triggering of the antigen by ANK61 antibodies activates the lytic machinery of rat NK cells, but not of T cells.
Target Species Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.