Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MS4A1, Mouse, Biotin, Clone: MEM 188, Abnova™

Catalog No. 89112612 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-612 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-612 Supplier Abnova Corporation Supplier No. MAB6000
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against NCAM1.

Specifications

Antigen MS4A1
Applications Flow Cytometry
Classification Monoclonal
Clone MEM 188
Conjugate Biotin
Description Mouse monoclonal antibody raised against NCAM1.
Dilution Flow Cytometry (10 ul/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene NCAM1
Gene Alias CD56/MSK39/NCAM
Gene Symbols NCAM1
Host Species Mouse
Immunogen KG-1 human acute myelogenous leukemia cell line.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4684
Test Specificity Specificity human CD56
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.