Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAM1, Mouse, Clone: 123C3, Abnova™

Catalog No. 89109465 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-465 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-465 Supplier Abnova Corporation Supplier No. MAB11301
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against NCAM1.

Specifications

Antigen NCAM1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 123C3.D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against NCAM1.
Dilution Western Blot (1:100-1:2000) Flow Cytometry (1-2 ug/106 cells) Immunocytochemistry/Immunofluorescence (1:10-1:2000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:200) Immunoprecipitation (1:10-1:500)
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene NCAM1
Gene Alias CD56/MSK39/NCAM
Gene Symbols NCAM1
Host Species Mouse
Immunogen A membrane preparation of a small cell lung carcinoma.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4684
Test Specificity This antibody recognizes two proteins (185 kDa & 145 kDa), identified as two isoforms of neural cell adhesion molecule (NCAM1/CD56).
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.