Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAM1, Mouse, FITC, Clone: MEM-188, Abnova™

Catalog No. 89112048 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-048 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-048 Supplier Abnova Corporation Supplier No. MAB5003
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native NCAM1.

Specifications

Antigen NCAM1
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-188
Conjugate FITC
Description Mouse monoclonal antibody raised against native NCAM1.
Dilution Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene NCAM1
Gene Alias CD56/MSK39/NCAM
Gene Symbols NCAM1
Host Species Mouse
Immunogen Native purified NCAM1 from KG-1 human acute myelogenous leukemia cell line.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4684
Test Specificity This antibody reacts with a 180 KDa isoform of CD56 (NCAM) expressed in leukocytes. It has been suggested that this antibody could react with rhesus monkey lymphocytes. Reactivity with other NCAM isoforms has not been tested.
Target Species Human, Primate
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.