Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAM2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104979 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-979 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-979 Supplier Abnova Corporation Supplier No. H00004685A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant NCAM2.

The protein encoded by this gene belongs to the immunoglobulin superfamily. It is a type I membrane protein and may function in selective fasciculation and zone-to-zone projection of the primary olfactory axons. [provided by RefSeq

Sequence: SIHGQPSSGKSFKLSITKQDDGGAPILEYIVKYRSKDKEDQWLEKKVQGNKDHIILEHLQWTMGYEVQITAANRLGYSEPTVYEFSMPPKPNIIKDTL

Specifications

Antigen NCAM2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant NCAM2.
Formulation 50% glycerol
Gene NCAM2
Gene Accession No. NM_004540
Gene Alias MGC51008/NCAM21
Gene Symbols NCAM2
Host Species Mouse
Immunogen NCAM2 (NP_004531, 598 a.a. ∽ 695 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4685
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.