Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAPG2 (LUZP5), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106993 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-993 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-993 Supplier Abnova Corporation Supplier No. H00054892A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant LUZP5.

Condensin complexes I and II play essential roles in mitotic chromosome assembly and segregation. Both condensins contain 2 invariant structural maintenance of chromosome (SMC) subunits, SMC2 (MIM 605576) and SMC4 (MIM 605575), but they contain different sets of non-SMC subunits. NCAPG2 is 1 of 3 non-SMC subunits that define condensin II (Ono et al., 2003 [PubMed 14532007]).[supplied by OMIM

Sequence: TKTGADVCRLWRIHQALYCFDYDLEESGEIKDMLLECFININYIKKEEGRRFLSCLFNWNINFIKMIHGTIKNQLQGLQKSLMVYIAEIYFRAWKKAS

Specifications

Antigen NCAPG2 (LUZP5)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant LUZP5.
Formulation 50% glycerol
Gene NCAPG2
Gene Accession No. NM_017760
Gene Alias CAP-G2/FLJ20311/LUZP5/MTB/hCAP-G2
Gene Symbols NCAPG2
Host Species Mouse
Immunogen LUZP5 (NP_060230, 177 a.a. ∽ 274 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54892
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.