Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCDN, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106560 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-560 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-560 Supplier Abnova Corporation Supplier No. H00023154A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant NCDN.

This gene encodes a leucine-rich cytoplasmic protein, which is highly similar to a mouse protein that negatively regulates Ca/calmodulin-dependent protein kinase II phosphorylation and may be essential for spatial learning processes. Several alternatively spliced transcript variants of this gene have been described. [provided by RefSeq

Sequence: NLACVEVRLALEETGTEVKEDVVTACYALMELGIQECTRCEQSLLKEPQKVQLVSVMKEAIGAVIHYLLQVGSEKQKEPFVFASVRILGAWLAEETSSLR

Specifications

Antigen NCDN
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant NCDN.
Formulation 50% glycerol
Gene NCDN
Gene Accession No. NM_001014839
Gene Alias KIAA0607
Gene Symbols NCDN
Host Species Mouse
Immunogen NCDN (NP_001014839, 312 a.a. ∽ 411 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23154
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.