Learn More
NCOR1, Mouse, Clone: 7A7A9, Abnova™
Mouse monoclonal antibody raised against partial recombinant NCOR1.
Supplier: Abnova Corporation MAB10376
Description
This gene encodes a protein that mediates ligand-independent transcription repression of thyroid-hormone and retinoic-acid receptors by promoting chromatin condensation and preventing access of the transcription machinery. It is part of a complex which also includes histone deacetylases and transcriptional regulators similar to the yeast protein Sin3p. This gene is located between the Charcot-Marie-Tooth and Smith-Magenis syndrome critical regions on chromosome 17. An alternatively spliced transcript variant has been described, but its full length sequence has not been determined. [provided by RefSeq
Specifications
NCOR1 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
NCOR1 | |
NCOR1 | |
Recombinant protein corresponding to amino acids 1-192 of human NCOR1. | |
RUO | |
9611 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Immunohistochemistry, Western Blot | |
7A7A9 | |
Mouse monoclonal antibody raised against partial recombinant NCOR1. | |
In ascites (0.03% sodium azide) | |
KIAA1047/MGC104216/N-CoR/TRAC1/hCIT529I10/hN-CoR | |
Mouse | |
100 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.