Learn More
NCOR1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant NCOR1.
Supplier: Abnova Corporation H00009611A01
Description
This gene encodes a protein that mediates ligand-independent transcription repression of thyroid-hormone and retinoic-acid receptors by promoting chromatin condensation and preventing access of the transcription machinery. It is part of a complex which also includes histone deacetylases and transcriptional regulators similar to the yeast protein Sin3p. This gene is located between the Charcot-Marie-Tooth and Smith-Magenis syndrome critical regions on chromosome 17. An alternatively spliced transcript variant has been described, but its full length sequence has not been determined. [provided by RefSeq
Sequence: GQGYLGTERPSSVSSVHSEGDYHRQTPGWAWEDRPSSTGSTQFPYNPLTMRMLSSTPPTPIACAPSAVNQAAPHQQNRIWEREPAPLLSAQYETLSDSDDSpecifications
NCOR1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant NCOR1. | |
NCOR1 | |
KIAA1047/MGC104216/N-CoR/TRAC1/hCIT529I10/hN-CoR | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_006311 | |
NCOR1 | |
NCOR1 (NP_006302, 2341 a.a. ∽ 2440 a.a) partial recombinant protein with GST tag. | |
RUO | |
9611 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only