Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCR1, Mouse, Clone: n1D9, Abnova™

Catalog No. 89108707 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-707 Supplier Abnova Corporation Supplier No. MAB1052
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant NCR1.

Specifications

Antigen NCR1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone n1D9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant NCR1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene NCR1
Gene Accession No. NM_004829
Gene Alias CD335/LY94/NK-p46/NKP46
Gene Symbols NCR1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 22-255 of human NCR1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9437
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.