Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDP52 Antibody (1813), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243588
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence
NDP52 Monoclonal specifically detects NDP52 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
NDP52 | |
Monoclonal | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
Antigen nuclear dot 52 kDa protein, calcium binding and coiled-coil domain 2, calcium-binding and coiled-coil domain-containing protein 2, NDP52MGC17318, Nuclear domain 10 protein 52, Nuclear domain 10 protein NDP52, Nuclear dot protein 52 | |
Mouse | |
52 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1813 | |
Unconjugated | |
PBS with No Preservative | |
CALCOCO2 | |
Recombinant protein encompassing a sequence within the center region of human NDP52. The exact sequence is proprietary. | |
Protein G purified | |
RUO | |
10241 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction