Learn More
Nectin-1 (CD111) Mouse anti-Human, PE, Clone: CK41, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 565766
Description
The CK41 monoclonal antibody specifically binds to Nectin-1, which is also known as, CD111, Poliovirus receptor-related protein 1(PRR1, PVRL1), Herpes virus entry mediator C (HveC), or Herpesvirus Ig-like receptor (HIgR). Nectin-1 is a type I transmembrane glycoprotein that belongs to the Ig gene superfamily. CD111 is expressed on a variety of cell types including, epithelial cells, endothelial cells, neuronal cells, megakaryocytes, and CD34 positive stem cells. CD111 functions as a receptor for herpes simplex virus 1 (HSV 1) and HSV 2. Nectin-1 also serves as an intercellular adhesion molecule helping to form junctions between cells, eg, between endothelial cells or epithelial cells. Nectin-1 can bind to other Nectin-1 molecules and Nectin family members and to CD155 (Poliovirus Receptor/PVR).

Specifications
Nectin-1 (CD111) | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Recombinant Protein | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
CK41 | |
Nectin1; PRR1; PVRL1; PVRR1; HveC; HIgR; CLPED1; ED4; SK-12; OFC7 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.