Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEDD9,, Mouse, Clone: C7.16D5.2G9.3C6, Abnova™

Catalog No. 89113664 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-664 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-664 Supplier Abnova Corporation Supplier No. MAB8969
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant NEDD9.

Specifications

Antigen NEDD9,
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone C7.16D5.2G9.3C6
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant NEDD9.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% sodium azide)
Gene NEDD9
Gene Alias CAS-L/CAS2/CASL/CASS2/HEF1/dJ49G10.2/dJ761I2.1
Gene Symbols NEDD9
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 82-398 of human NEDD9.
Purification Method Purified ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4739
Test Specificity This antibody is specific only for HEF1 and does not recognize p130Cas. Have not tested on Sin1.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.