Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NEFH, Mouse, Clone: NAP4, Abnova™
Mouse monoclonal antibody raised against native NEFH.
Supplier: Abnova Corporation MAB2320
Specifications
NEFH | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) Western Blot (1:5000) The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μL | |
Primary | |
Avian, Human, Mammalia | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
NAP4 | |
Mouse monoclonal antibody raised against native NEFH. | |
In 10mM sodium phosphate buffer, 250mM NaCl, pH 7.6 (15mg/mL BSA, 5mM sodium azide, 0.01% thimerosal) | |
Native purified neurofilaments from pig spinal cord. | |
RUO | |
Specifically recognizes the phosphorylated variant of NF-H subunit (∼180-220 KDa), showing some weaker reactivity with phosphorylated forms of NF-M. | |
Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction