Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEFL, Mouse, Clone: DA2, Abnova™

Catalog No. 89110929 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-929 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-929 Supplier Abnova Corporation Supplier No. MAB2321
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native NEFL.

Specifications

Antigen NEFL
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone DA2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native NEFL.
Dilution Immunohistochemistry (1:250) Immunofluorescence (1:100-1:500) Western Blot (1:5000-1:10000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene LOC733642
Gene Symbols LOC733642
Host Species Mouse
Immunogen Native purified pig NEFL.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 733642
Test Specificity Specifically recognizes the light neurofilament subunit or NF-L (∼68 KDa).
Target Species Avian, Human, Mammalia
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.