Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEUROD6, Mouse, Clone: 4G11, Abnova™

Catalog No. 89107262 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-262 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-262 Supplier Abnova Corporation Supplier No. H00063974M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant NEUROD6.

NEUROD6 is a member of the NEUROD (NEUROD1; MIM 601724) family of basic helix-loop-helix (bHLH) transcription factors (Guo et al., 2002 [PubMed 12357074]).[supplied by OMIM

Sequence: GQGGEAAHHTRSPYSTFYPPYHSPELTTPPGHGTLDNSKSMKPYNYCSAYESFYESTSPECASPQFEGPLSPPPINYNGIFSLKQEETLDYGKNYNYGMH

Specifications

Antigen NEUROD6
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4G11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant NEUROD6.
Formulation ascites with no preservative
Gene NEUROD6
Gene Accession No. NM_022728
Gene Alias Atoh2/MATH2/Math-2/NEX1M/bHLHa2
Gene Symbols NEUROD6
Host Species Mouse
Immunogen NEUROD6 (NP_073565.2,191 a.a. ∽ 290 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63974
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.