Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofilament, Mouse, Clone: 403, Abnova™

Catalog No. 89135909 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-909 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-909 Supplier Abnova Corporation Supplier No. MAB5186
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Neurofilament.

Specifications

Antigen Neurofilament
Applications Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 403
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Neurofilament.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10) The optimal working dilution should be determined by the end user.
Formulation In tissue culture supernatant (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified human Neurofilament.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The monoclonal antibody reacts specific with the medium component of the three subunits of neurofilaments. In immunoblots it reacts with the 160 kD protein. Neurofilaments, the intermediate filaments of neurons, are composed of three polypeptides with a moleculeweight of respecitively, 70, 160 and 200 kD. The three polypeptides can be differentially expressed during neuronal development, or in adult brain tissue. Antibodies directed against the three different neurofilament proteins can be an important aid in studying neurofilament expression pattern and in testing of tumours.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.