Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neurofilament heavy protein, Mouse, Clone: NF-01, Abnova™

Catalog No. 89135484 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-484 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-484 Supplier Abnova Corporation Supplier No. MAB3638
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Neurofilament heavy protein .

Specifications

Antigen Neurofilament heavy protein
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone NF-01
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Neurofilament heavy protein .
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified porcine Neurofilament heavy protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes a phosphorylated epitope on heavy neurofilament protein (210 KDa) of various species. Antibodies to the various neurofilament subunits are very useful cell type markers since the proteins are among the most abundant of the nervous system, are expressed only in neurons and are biochemically very stable.
Target Species Mammalia, Pig
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.