Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NF1, Mouse, Clone: McNFn27, Abnova™

Catalog No. 89110920 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-920 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-920 Supplier Abnova Corporation Supplier No. MAB2306
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of NF1.

This gene product appears to function as a negative regulator of the ras signal transduction pathway. Mutations in this gene have been linked to neurofibromatosis type 1, juvenile myelomonocytic leukemia and Watson syndrome. The mRNA for this gene is subject to RNA editing (CGA>UGA->Arg1306Term) resulting in premature translation termination. Alternatively spliced transcript variants encoding different isoforms have also been described for this gene. [provided by RefSeq

Specifications

Antigen NF1
Applications ELISA, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone McNFn27
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of NF1.
Dilution Immunohistochemistry (10-20 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene NF1
Gene Alias DKFZp686J1293/FLJ21220/NFNS/VRNF/WSS
Gene Symbols NF1
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 27-41 of human NF1.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4763
Target Species Guinea Pig, Human, Mouse, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.