Learn More
NF1, Mouse, Clone: McNFn27a, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of NF1.
Supplier: Abnova Corporation MAB2305
Description
This gene product appears to function as a negative regulator of the ras signal transduction pathway. Mutations in this gene have been linked to neurofibromatosis type 1, juvenile myelomonocytic leukemia and Watson syndrome. The mRNA for this gene is subject to RNA editing (CGA>UGA->Arg1306Term) resulting in premature translation termination. Alternatively spliced transcript variants encoding different isoforms have also been described for this gene. [provided by RefSeq
Specifications
NF1 | |
Monoclonal | |
Unconjugated | |
Western Blot (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
NF1 | |
NF1 | |
A synthetic peptide corresponding to amino acids 27-41 of human NF1. | |
RUO | |
4763 | |
Guinea Pig, Human, Mouse, Rat | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
McNFn27a | |
Mouse monoclonal antibody raised against synthetic peptide of NF1. | |
In PBS (0.05% sodium azide) | |
DKFZp686J1293/FLJ21220/NFNS/VRNF/WSS | |
Mouse | |
200 μL | |
Primary | |
This antibody is specific to full-length or truncated forms of neurofibromin. | |
Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.