Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NF200, Mouse, Clone: NF-200, Abnova™
Mouse monoclonal antibody raised against native NF200.
Supplier: Abnova Corporation MAB1523
Specifications
NF200 | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Immunohistochemistry (Frozen sections) (1-2 ug/mL) Western Blot (0.5 mL) The optimal working dilution should be determined by the end user. | |
Mouse | |
Affinity purification | |
RUO | |
Human, Mouse, Pig, Rat | |
Lyophilized |
Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot | |
N52 | |
Mouse monoclonal antibody raised against native Nefh. | |
Lyophilized from 1.2% sodium acetate (2mg BSA, 0.01mg sodium azide) | |
Native purified pig neurofilament 200. | |
100 μg | |
Primary | |
Store at -20°C on dry atmosphere. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction