Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFIA, Mouse, Clone: 1E11, Abnova™

Catalog No. 89104987 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-987 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-987 Supplier Abnova Corporation Supplier No. H00004774M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant NFIA.

Nuclear factor I (NFI) proteins constitute a family of dimeric DNA-binding proteins with similar, and possibly identical, DNA-binding specificity. They function as cellular transcription factors and as replication factors for adenovirus DNA replication. Diversity in this protein family is generated by multiple genes, differential splicing, and heterodimerization.[supplied by OMIM

Sequence: MYSPLCLTQDEFHPFIEALLPHVRAFAYTWFNLQARKRKYFKKHEKRMSKEEERAVKDELLSEKPEVKQKWASRLLAKLRKDIRPEYREDFVLTVTGKKPPCCVLSNPDQKGKMRRIDCLRQADKVWRLDLVMVILFKGIPLESTDGERLVKSPQCSNPGLCVQPHHIGVSVKELDLYLAYFVHAADSSQSESPSQPSDADIKDQPENGHLGFQDSFVTSGVFSVTELVRVSQTPIAAGTGPNFSLSDLESSSYYSMSPGAMRRSLPSTSSTSSTKRLKSVEDEMDSPGEEPFYTGQGRSPGSGSQSSGWHEVEPGMPSPTTLKKSEKSGFSSPSPSQTSSLGTAFTQHHRPVITGPRASPHATPSTLHFPTSPIIQQPGPYFSHPAIRYHPQETLKEFVQLVCPDAGQQAGQVGFLNPNGSSQGKVHNPFLPTPMLPPPPPPPMARPVPLPVPDTKPPTTSTEGGAASPTSPILVPGIKVAASHHPPDRPPDPFSTL

Specifications

Antigen NFIA
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant NFIA.
Formulation ascites with no preservative
Gene NFIA
Gene Accession No. BC022264
Gene Alias DKFZp434L0422/KIAA1439/NFI-L
Gene Symbols NFIA
Host Species Mouse
Immunogen NFIA (AAH22264,1 a.a. ∽ 498 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4774
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.