Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NFIC, Mouse, Clone: 2B6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant NFIC.
Supplier: Abnova Corporation H00004782M04A
Description
Sequence: TEDMEGGISSPVKKTEMDKSPFNSPSPQDSPRLSSFTQHHRPVIAVHSGIARSPHPSSALHFPTTSILPQTASTYFPHTAIRYPPHLNPQDPLKDLVSLACDPASQQPGPSpecifications
| NFIC | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_005597 | |
| NFIC | |
| NFIC (NP_005588,314 a.a. ∽ 423 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4782 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 2B6 | |
| Mouse monoclonal antibody raised against a partial recombinant NFIC. | |
| NFIC | |
| CTF/CTF5/MGC20153/NF-I/NFI | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction