Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFYB, Mouse, Clone: 2H2, Abnova™

Catalog No. 89104999 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-999 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-999 Supplier Abnova Corporation Supplier No. H00004801M08A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant NFYB.

The protein encoded by this gene is one subunit of a trimeric complex, forming a highly conserved transcription factor that binds with high specificity to CCAAT motifs in the promoter regions in a variety of genes. This gene product, subunit B, forms a tight dimer with the C subunit, a prerequisite for subunit A association. The resulting trimer binds to DNA with high specificity and affinity. Subunits B and C each contain a histone-like motif. Observation of the histone nature of these subunits is supported by two types of evidence; protein sequence alignments and experiments with mutants. [provided by RefSeq

Sequence: MTMDGDSSTTDASQLGISADYIGGSHYVIQPHDDTEDSMNDHEDTNGSKESFREQDIYLPIANVARIMKNAIPQTGKIAKDAKECVQECVSEFISFITSEASERCHQEKRKTINGEDILFAMSTLGFDSYVEPLKLYLQKFREAMKGEKGIGGAVTATDGLSEELTEEAFTNQLPAGLITTDGQQQNVMVYTTSYQQISGVQQIQFS

Specifications

Antigen NFYB
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2H2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant NFYB.
Formulation ascites with no preservative
Gene NFYB
Gene Accession No. BC005317
Gene Alias CBF-A/CBF-B/HAP3/NF-YB
Gene Symbols NFYB
Host Species Mouse
Immunogen NFYB (AAH05317,1 a.a. ∽ 207 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4801
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.