Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NFYC, Mouse, Clone: 1D3, Abnova™

Catalog No. 89105000 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-000 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-000 Supplier Abnova Corporation Supplier No. H00004802M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant NFYC.

This gene encodes one subunit of a trimeric complex forming a highly conserved transcription factor that binds with high specificity to CCAAT motifs in the promoters of a variety of genes. The encoded protein, subunit C, forms a tight dimer with the B subunit, a prerequisite for subunit A association. The resulting trimer binds to DNA with high specificity and affinity. Subunits B and C each contain a histone-like motif. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: DAQQSLQSFWPRVMEEIRNLTVKDFRVQELPLARIKKIMKLDEDVKMISAEAPVLFAKAAQIFITELTLRAWIHTEDNKRRTLQRNDIAMAITKFDQFDF

Specifications

Antigen NFYC
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant NFYC.
Formulation ascites with no preservative
Gene NFYC
Gene Accession No. BC005003.1
Gene Alias CBF-C/CBFC/DKFZp667G242/FLJ45775/H1TF2A/HAP5/HSM/NF-YC
Gene Symbols NFYC
Host Species Mouse
Immunogen NFYC (AAH05003.1,14 a.a. ∽ 113 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4802
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.