Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NGFR, Mouse, Clone: 2F1C2, Abnova™

Catalog No. 89109035 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-035 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-035 Supplier Abnova Corporation Supplier No. MAB10831
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant NGFR.

Nerve growth factor receptor contains an extracellular domain containing four 40-amino acid repeats with 6 cysteine residues at conserved positions followed by a serine/threonine-rich region, a single transmembrane domain, and a 155-amino acid cytoplasmic domain. The cysteine-rich region contains the nerve growth factor binding domain. [provided by RefSeq

Specifications

Antigen NGFR
Applications ELISA, Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2F1C2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant NGFR.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene NGFR
Gene Alias CD271/Gp80-LNGFR/TNFRSF16/p75(NTR)/p75NTR
Gene Symbols NGFR
Host Species Mouse
Immunogen Recombinant protein corresponding to human NGFR.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4804
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.