Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ngfr, Mouse, Clone: MC192, Abnova™

Catalog No. 89112305 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-305 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-305 Supplier Abnova Corporation Supplier No. MAB5595
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Ngfr.

Specifications

Antigen Ngfr
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone MC192
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Ngfr.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from cell culture supernatant (1% BSA)
Gene Ngfr
Gene Alias LNGFR/RNNGFRR/p75/p75NTR
Gene Symbols Ngfr
Host Species Mouse
Immunogen Rat Ngfr.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24596
Test Specificity Specificity has been confirmed by immunohistochemical staining of lesioned sciatic nerve and spinal cord, the results reflect the reported literatures.
Target Species Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 100μL sterilzed water store at -20°C or lower. For an additional stability glycerol (1:1) may be added.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.