Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ngfr, Mouse, Clone: ME20.4, Abnova™

Catalog No. 89112306 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-306 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-306 Supplier Abnova Corporation Supplier No. MAB5596
Add to Cart
Add to Cart

Mouse (P75 knock-out) monoclonal antibody raised against native NGFR.

Nerve growth factor receptor contains an extracellular domain containing four 40-amino acid repeats with 6 cysteine residues at conserved positions followed by a serine/threonine-rich region, a single transmembrane domain, and a 155-amino acid cytoplasmic domain. The cysteine-rich region contains the nerve growth factor binding domain. [provided by RefSeq

Specifications

Antigen Ngfr
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone ME20.4
Conjugate Unconjugated
Description Mouse (P75 knock-out) monoclonal antibody raised against native NGFR.
Dilution Immunohistochemistry (2 ug/mL) Flow Cytometry (20 ug/mL) ELISA (1:5000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene NGFR
Gene Alias CD271/Gp80-LNGFR/TNFRSF16/p75(NTR)/p75NTR
Gene Symbols NGFR
Host Species Mouse
Immunogen Native from human WM245 melanoma cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4804
Test Specificity This antibody recognizes p75NTR (low affinity neurotrophin receptor).
Target Species Canine, Feline, Human, Pig, Rabbit, Sheep
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.