Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX3-1, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89110918 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-918 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-918 Supplier Abnova Corporation Supplier No. MAB2304
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant NKX3-1.

The homeodomain-containing transcription factor NKX3-1 is a putative prostate tumor suppressor that is expressed in a largely prostate-specific and androgen-regulated manner. Loss of NKX3-1 protein expression is a common finding in human prostate carcinomas and prostatic intraepithelial neoplasia.[supplied by OMIM

Specifications

Antigen NKX3-1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant NKX3-1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene NKX3-1
Gene Alias BAPX2/NKX3/NKX3.1/NKX3A
Gene Symbols NKX3-1
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to full length human NKX3-1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4824
Target Species Mouse
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.