Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKX3-2 (BAPX1), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104140 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-140 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-140 Supplier Abnova Corporation Supplier No. H00000579A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant BAPX1.

This gene encodes a member of the NK family of homeobox-containing proteins. The encoded protein may play a role in skeletal development. [provided by RefSeq

Sequence: RLFGERDAGALGGAEDSLLASPAGTRTAAGRTAESPEGWDSDSALSEENESRRRCADAR

Specifications

Antigen NKX3-2 (BAPX1)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant BAPX1.
Formulation 50% glycerol
Gene NKX3-2
Gene Accession No. NM_001189
Gene Alias BAPX1/MGC138171/NKX3.2/NKX3B
Gene Symbols NKX3-2
Host Species Mouse
Immunogen BAPX1 (NP_001180, 53 a.a. ∽ 111 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 579
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.