Learn More
NLRC4 (CARD12), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CARD12.
Supplier: Abnova Corporation H00058484A01
Description
In C. elegans, Ced4 binds and activates Ced3, an apoptotic initiator caspase, via caspase-associated recruitment domains (CARDs). Human Ced4 homologs include APAF1 (MIM 602233), NOD1/CARD4 (MIM 605980), and NOD2/CARD15 (MIM 605956). These proteins have at least 1 N-terminal CARD domain followed by a centrally located nucleotide-binding domain (NBD or NACHT) and a C-terminal regulatory domain, found only in mammals, that contains either WD40 repeats or leucine-rich repeats (LRRs). CARD12 is a member of the Ced4 family and can induce apoptosis.[supplied by OMIM
Sequence: QESLQSVKNTTEQEILKAININSFVECGIHLYQESTSKSALSQEFEAFFQGKSLYINSGNIPDYLFDFFEHLPNCASALDFIKLDFYGGAMASWEKAAEDSpecifications
NLRC4 (CARD12) | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant CARD12. | |
NLRC4 | |
CARD12/CLAN/CLAN1/CLANA/CLANB/CLANC/CLAND/CLR2.1/IPAF | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC031555 | |
NLRC4 | |
CARD12 (AAH31555, 531 a.a. ∽ 630 a.a) partial recombinant protein with GST tag. | |
RUO | |
58484 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only